Learn More
Marlin Steel Wire Products Mesh Basket with Handles 15"L x 9"W x 6.50"H
SDP
Catalog No. NC1959489
Encompass_Preferred
Rectangular material handling basket with handles for easy gripping.Stacks so that parts inside baskets do not get crushedrobust quality that is ideal for washing parts, degreasing, ultrasonic cleaning, curing, and material handling in a variety of applications such as chemical bath, heat treating, cryogenics, and food processing4 mesh openings per linear inch
Catalog No. NC1959489 Supplier Marlin Steel Wire Products Supplier No. 000036818838
Rectangular material handling basket with handles for easy gripping.Stacks so that parts inside baskets do not get crushedrobust quality that is ideal for washing parts, degreasing, ultrasonic cleaning, curing, and material handling in a variety of applications such as chemical bath, heat treating, cryogenics, and food processing4 mesh openings per linear inch