Learn More
Marlin Steel Wire Products Mesh Basket with Lid 5"L x 3"W x 1"H
SDP
Catalog No. NC1959503
Encompass_Preferred
Round material handling basket with handles for easy gripping.robust quality that is ideal for washing parts, degreasing, ultrasonic cleaning, curing, and material handling in a variety of applications such as chemical bath, heat treating, cryogenics, and food processing24 mesh openings per linear inch
Catalog No. NC1959503 Supplier Marlin Steel Wire Products Supplier No. 000036821338
Round material handling basket with handles for easy gripping.robust quality that is ideal for washing parts, degreasing, ultrasonic cleaning, curing, and material handling in a variety of applications such as chemical bath, heat treating, cryogenics, and food processing24 mesh openings per linear inch