Learn More
Marlin Steel Wire Products Mesh Basket with Lid 9"L x 7"W x 3"H
SDP
Supplier:  Marlin Steel Wire Products 000036822138
Encompass_Preferred
Stainless steel material handling basket with a lid and hinge.10 mesh openings per linear inchUsed for small material handling or washing. Rugged construction ensures basket longevity in applications such as chemical baths, heat treating, cryogenics, and food processing.
Catalog No. NC1959530