Learn More
Electron Microscopy Sciences Tissue-Tek Mesh Biopsy Cassette, Pink
SDP
Catalog No. 5029453
Encompass
Tissue-Tek Mesh Biopsy Cassette System is now the easier, ideal way for processing the smallest of biopsy specimens. Using Tissue-Tek Mesh Biopsy Cassettes, you avoid reagent carryover in the tissue processor and you eliminate the structural artifacts usually caused by sponge and tea bag abrasion. They also allow direct specimen filtration into the cassette and facilitate embedding.
Flow-through improved by large venting slots.
Easy visualization and safe specimen containment facilitated by a mesh screen with blue tint.
Tongue-and-groove seal sorrounds entire specimen.
Rounded inner walls allow for easier removal.
Color: Pink
Model: 4307
Catalog No. 50-294-53 Supplier Electron Microscopy Sciences Supplier No. 62502P
Tissue-Tek Mesh Biopsy Cassette System is now the easier, ideal way for processing the smallest of biopsy specimens. Using Tissue-Tek Mesh Biopsy Cassettes, you avoid reagent carryover in the tissue processor and you eliminate the structural artifacts usually caused by sponge and tea bag abrasion. They also allow direct specimen filtration into the cassette and facilitate embedding.
Flow-through improved by large venting slots.
Easy visualization and safe specimen containment facilitated by a mesh screen with blue tint.
Tongue-and-groove seal sorrounds entire specimen.
Rounded inner walls allow for easier removal.
Color: Pink
Model: 4307