Learn More
Grainger Mesh Panel Retainer Anodized Aluminum
Catalog No. 502764038
Encompass_Preferred
The 80/20 Mesh Panel Retainer for Mesh Panels is a metal retainer that holds mesh panels in place on 80/20 aluminum framing. It is made from 6061-T6 aluminum and has a black anodized finish. The retainer measures 0.625" wide x 0.625" tall x 1.625" long. It has pre-drilled holes that allow it to be mounted onto 80/20 framing using fasteners (not included). This versatile retainer securely holds mesh panels onto 80/20 frames for enclosures, machine guards, partitions and more. It is sold individually.
Catalog No. 50-276-4038 Supplier Grainger Supplier No. 16U366
The 80/20 Mesh Panel Retainer for Mesh Panels is a metal retainer that holds mesh panels in place on 80/20 aluminum framing. It is made from 6061-T6 aluminum and has a black anodized finish. The retainer measures 0.625" wide x 0.625" tall x 1.625" long. It has pre-drilled holes that allow it to be mounted onto 80/20 framing using fasteners (not included). This versatile retainer securely holds mesh panels onto 80/20 frames for enclosures, machine guards, partitions and more. It is sold individually.