Learn More
Grainger Mesh Panel Retainer Anodized Aluminum
Catalog No. 502766618
Encompass_Preferred
The 16U349 Mesh Panel Retainer is an anodized aluminum retainer designed to hold mesh panels in place. It measures 0.5" wide x 0.25" thick and comes in lengths of 36", 48", 60", and 72". The retainer has a slot that is 0.0625" wide x 0.1875" deep to accept 1/16" thick mesh panels. It can be used to mount mesh panels onto electronic enclosures, machine guards, partitions and more. The anodized aluminum provides corrosion resistance. Overall, this product provides an easy way to securely mount mesh panels for ventilation, safety guards, or decorative purposes.
Catalog No. 50-276-6618 Supplier Grainger Supplier No. 16U349
The 16U349 Mesh Panel Retainer is an anodized aluminum retainer designed to hold mesh panels in place. It measures 0.5" wide x 0.25" thick and comes in lengths of 36", 48", 60", and 72". The retainer has a slot that is 0.0625" wide x 0.1875" deep to accept 1/16" thick mesh panels. It can be used to mount mesh panels onto electronic enclosures, machine guards, partitions and more. The anodized aluminum provides corrosion resistance. Overall, this product provides an easy way to securely mount mesh panels for ventilation, safety guards, or decorative purposes.