Learn More
Grainger Mesh Panel Retainer Anodized Aluminum
Catalog No. 502766751
Encompass_Preferred
The 80/20 Panel Retainer for Standard Panel is a metal panel retainer designed to hold standard 80/20 panels in place. It is made from aluminum and has a black anodized finish. The panel retainer measures 0.31" x 0.31" x 0.75" and weighs 0.01 lbs. It can accommodate panel thicknesses from 0.25" to 0.375" and attaches to 80/20 T-slot aluminum profiles using two 1/4"-20 socket head cap screws (sold separately). This panel retainer securely holds panels to 80/20 framing without drilling or tapping. It is easy to install and provides a clean look.
Catalog No. 50-276-6751 Supplier Grainger Supplier No. 16U352
The 80/20 Panel Retainer for Standard Panel is a metal panel retainer designed to hold standard 80/20 panels in place. It is made from aluminum and has a black anodized finish. The panel retainer measures 0.31" x 0.31" x 0.75" and weighs 0.01 lbs. It can accommodate panel thicknesses from 0.25" to 0.375" and attaches to 80/20 T-slot aluminum profiles using two 1/4"-20 socket head cap screws (sold separately). This panel retainer securely holds panels to 80/20 framing without drilling or tapping. It is easy to install and provides a clean look.