Learn More
Grainger Mesh Panel Retainer Anodized Aluminum
Catalog No. 502766655
Encompass_Preferred
The 80/20 Mesh Panel Retainer for Mesh Panels is a metal retainer that holds mesh panels in place on 80/20 aluminum framing. It is made from 6061-T6 aluminum and has a black anodized finish. The retainer measures 0.625" wide x 0.375" tall and has two set screws to lock it to the 80/20 framing. It is designed to fit 1" series 80/20 framing and hold mesh panels up to 0.25" thick in place. This high quality retainer provides an easy way to mount mesh panels for enclosures, machine guards, partitions and more using 80/20 aluminum extrusion frames.
Catalog No. 50-276-6655 Supplier Grainger Supplier No. 16U353
The 80/20 Mesh Panel Retainer for Mesh Panels is a metal retainer that holds mesh panels in place on 80/20 aluminum framing. It is made from 6061-T6 aluminum and has a black anodized finish. The retainer measures 0.625" wide x 0.375" tall and has two set screws to lock it to the 80/20 framing. It is designed to fit 1" series 80/20 framing and hold mesh panels up to 0.25" thick in place. This high quality retainer provides an easy way to mount mesh panels for enclosures, machine guards, partitions and more using 80/20 aluminum extrusion frames.