Learn More
Kishigo Mesh Pants
Catalog No. 19220927
Encompass
Change view
Click to view available options
Size:
2X-Large/3X-Large
Large/X-Large
Color:
Orange
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19-220-927 Large/X-Large Orange
19-220-926 2X-Large/3X-Large Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-220-927 Supplier Kishigo Supplier No. 3107LXL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ultra-Cool™ 100% polyester mesh pants
  • Ultra-Cool™ 100% polyester mesh
  • Drawstring waist with elastic back
  • Elastic bottom cuffs
  • 2 in. wide reflective material
  • Left and right side pocket access slits
Compliance
Meets ANSI™ 107 Class E standards.
Specifications
Size Large/X-Large
Gender Unisex
Product Type Hi-vis Mesh Jogger
Material Polyester
Color Orange
No. of Pockets 2
Construction 5-needle overlap seam
Certifications/Compliance ANSI 107 Class 3, Class E
Style Elastic bottom cuffs