Learn More
Med Vet International Premilene Mesh 26cm x 36cm, Sterile, J1249C, Each
Catalog No. 5000495661
Encompass_Preferred
Premilene Mesh is thin, soft, smooth and strong. The mesh of choice for larger hernia repair. • Made from Polypropylene Monofilament, nonabsorbable • Smaller pore size • Can be trimmed with no fraying • Thin mesh: transparent, can see underlying tissues • Suitable for minimally invasive techniques Specifications: Knitted Weight 82g/m2 Thickness 0.48mm Pore Size 0.8mm Premilene Mesh 26.0cm x 36.0cm
Catalog No. 50-004-95661 Supplier Med Vet International Supplier No. JOR1249C
Premilene Mesh is thin, soft, smooth and strong. The mesh of choice for larger hernia repair. • Made from Polypropylene Monofilament, nonabsorbable • Smaller pore size • Can be trimmed with no fraying • Thin mesh: transparent, can see underlying tissues • Suitable for minimally invasive techniques Specifications: Knitted Weight 82g/m2 Thickness 0.48mm Pore Size 0.8mm Premilene Mesh 26.0cm x 36.0cm