Learn More
Med Vet International Optilene Surgical Mesh 5cm x 10cm, Sterile, J1247A, Each
Catalog No. 5000495740
Encompass_Preferred
• Monofilament Polypropylene nonabsorbable • Larger pore size • Blue guidelines in material helps proper placement • More elastic mesh • Can be used on minimally invasive surgeries Specifications: Weight 60g/m2 Thickness 0.53mm Pore Size 1.5mm Optilene Mesh 5.0cm x 10.0cm
Catalog No. 50-004-95740 Supplier Med Vet International Supplier No. JOR1247A
• Monofilament Polypropylene nonabsorbable • Larger pore size • Blue guidelines in material helps proper placement • More elastic mesh • Can be used on minimally invasive surgeries Specifications: Weight 60g/m2 Thickness 0.53mm Pore Size 1.5mm Optilene Mesh 5.0cm x 10.0cm