Learn More
Grainger Buyers Products Manual Dump Tarp System, PVC-Coated Vinyl Mesh, 6.5 ft x 18 ft, 1 ea
Catalog No. 502785139
Encompass_Preferred
This manual dump truck and trailer tarp system is designed for simple dump body and dump trailer applications, providing a reliable covering solution. The kit features a durable extruded aluminum tarp roller that telescopes to the exact width of your vehicle for a precise fit. The heavy-duty PVC-coated vinyl mesh tarp includes brass grommets and reinforced, double-stitched hems to ensure durability in outdoor environments.
  • Features a 7 in. folding crank with detent for manual operation
  • Includes a standard sewn-in rear pocket to fit an optional anti-sail retention bow
  • Tarp material is UV resistant for extended service life
  • Supplied with a 20 ft. pull rope, mounting brackets, rope tie-down hooks, and two rubber tarp straps with S-hooks
  • Designed for ease of installation with all necessary mounting hardware included
Catalog No. 50-278-5139 Supplier Grainger Supplier No. 19A806
This manual dump truck and trailer tarp system is designed for simple dump body and dump trailer applications, providing a reliable covering solution. The kit features a durable extruded aluminum tarp roller that telescopes to the exact width of your vehicle for a precise fit. The heavy-duty PVC-coated vinyl mesh tarp includes brass grommets and reinforced, double-stitched hems to ensure durability in outdoor environments.
  • Features a 7 in. folding crank with detent for manual operation
  • Includes a standard sewn-in rear pocket to fit an optional anti-sail retention bow
  • Tarp material is UV resistant for extended service life
  • Supplied with a 20 ft. pull rope, mounting brackets, rope tie-down hooks, and two rubber tarp straps with S-hooks
  • Designed for ease of installation with all necessary mounting hardware included