Learn More
Grainger Buyers Products Dump Tarp System, PVC-Coated Vinyl Mesh, 6 ft. x 14 ft., 1 ea.
Catalog No. 502785397
Encompass_Preferred
This manual dump truck and trailer tarp system is designed for simple, reliable load coverage on dump bodies and dump trailers. The kit includes a durable 6 ft. x 14 ft. PVC-coated vinyl mesh tarp and all necessary mounting hardware to facilitate quick installation and operation.
  • Features extruded aluminum cross bars that telescope to match the exact bed width of dumps and trailers up to 102 in. wide
  • Constructed with heavy-duty, double-stitched reinforced hems and brass grommets for added durability
  • Includes a 7 in. folding crank with detent, tarp roller, 20 ft. pull rope, rope tie-down hooks, mounting brackets, and two rubber tarp straps with S-hooks
  • Utilizes UV-resistant mesh material for long-term outdoor performance
  • Comes with a standard sewn-in rear pocket to accommodate an optional anti-sail retention bow
Catalog No. 50-278-5397 Supplier Grainger Supplier No. 19A807
This manual dump truck and trailer tarp system is designed for simple, reliable load coverage on dump bodies and dump trailers. The kit includes a durable 6 ft. x 14 ft. PVC-coated vinyl mesh tarp and all necessary mounting hardware to facilitate quick installation and operation.
  • Features extruded aluminum cross bars that telescope to match the exact bed width of dumps and trailers up to 102 in. wide
  • Constructed with heavy-duty, double-stitched reinforced hems and brass grommets for added durability
  • Includes a 7 in. folding crank with detent, tarp roller, 20 ft. pull rope, rope tie-down hooks, mounting brackets, and two rubber tarp straps with S-hooks
  • Utilizes UV-resistant mesh material for long-term outdoor performance
  • Comes with a standard sewn-in rear pocket to accommodate an optional anti-sail retention bow