Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Mesh Water Recovery Body Bags
SDP

Catalog No. 22446185 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W):
96 x 38 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-446-185 96 x 38 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-446-185 Supplier Mopec Supplier No. FS076
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Durable, high visibility mesh body bags provide efficient water recovery with rapid drainage and convenient handling.

  • Built for reliability in demanding conditions, this set includes three bags, each with an identification tag kit.
  • Features a Clam-shell zipper and 10 carry straps for convenient handling.
  • Made from High-visibility orange, Vinyl-coated polyester scrim mesh.
  • Measures (96 L x 38 W) in.
  • Drains quickly when removed from water.
  • Supplied 3 per case and includes 3 ID tags.

Specifications

Dimensions (L x W) 96 x 38 in.
Product Type Body Bag
Color Orange
Quantity 3/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.