Learn More
Electron Microscopy Sciences MeshWell Plate 12 well
SDP
Catalog No. 5030872
Encompass
These plates are made from polypropylene and the bottom of each of the wells is an ETFE monofilament screening fabric with a 500um (0.0197") opening. Use with Reagent Trays (64730 - 64732). Comes with one plate and two trays.
Catalog No. 50-308-72 Supplier Electron Microscopy Sciences Supplier No. 6470012
These plates are made from polypropylene and the bottom of each of the wells is an ETFE monofilament screening fabric with a 500um (0.0197") opening. Use with Reagent Trays (64730 - 64732). Comes with one plate and two trays.