Learn More
Electron Microscopy Sciences MeshWell Plate 24 well
SDP
Supplier:  Electron Microscopy Sciences 6470024
Encompass
These plates are made from polypropylene and the bottom of each of the wells is an ETFE monofilament screening fabric with a 500um (0.0197") opening. Use with Reagent Trays (64730 - 64732). Comes with one plate and two trays.
Catalog No. 50-308-73