Learn More
Electron Microscopy Sciences MeshWell Plate 24 with Tray and Lid
SDP
Catalog No. 501927773
Encompass_Preferred
These plates are made from polypropylene and the bottom of each of the wells is an ETFE monofilament screening fabric with a 500um (0.0197") opening.
Catalog No. 50-192-7773 Supplier Electron Microscopy Sciences Supplier No. 6470301
These plates are made from polypropylene and the bottom of each of the wells is an ETFE monofilament screening fabric with a 500um (0.0197") opening.