Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mesitaldehyde, 97%

Catalog No. p-7080539
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1010404 2 g
AAA1010409 10 g
AAA1010418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1010404 Supplier Thermo Scientific Chemicals Supplier No. A1010404
Add to Cart

CAS: 487-68-3 | C10H12O | 148.21 g/mol

Mesitaldehyde is used in organic synthesis, and also used as medicine and other fine chemical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mesitaldehyde is used in organic synthesis, and also used as medicine and other fine chemical products.

Solubility
Soluble in Chloroform

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 487-68-3
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00003341
InChI Key HIKRJHFHGKZKRI-UHFFFAOYSA-N
Synonym mesitaldehyde, mesitylaldehyde, benzaldehyde, 2,4,6-trimethyl, 2-mesitylenecarboxaldehyde, mesitylenecarboxaldehyde, 2-formylmesitylene, 2,4,6-trimethyl-benzaldehyde, isodural, mesityl aldehyde, b-isodural
PubChem CID 10254
IUPAC Name 2,4,6-trimethylbenzaldehyde
SMILES CC1=CC(C)=C(C=O)C(C)=C1

Specifications

Melting Point 14°C
Density 1.005
Boiling Point 237°C
Flash Point 106°C (222°F)
Refractive Index 1.554
Quantity 2 g
Beilstein 1364114
Sensitivity Air Sensitive
Solubility Information Soluble in Chloroform
Formula Weight 148.21
Percent Purity 98%
Chemical Name or Material Mesitaldehyde
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-662-1
RTECSNumber CU8500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.