Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Mesitylene

Catalog No. M72005ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
10 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M72005ML 5 mL
M720010L 10 L
M7200500ML 500 mL
M7200100ML 100 mL
M720025L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. M72005ML Supplier Sigma Aldrich Supplier No. M72005ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Mesitylene

Specifications

CAS 108-67-8
Melting Point -45°C
Density 0.864 g/mL at 25°C
Boiling Point 163°C to 166°C
Molecular Formula C9H12
Refractive Index n20/D 1.499
Linear Formula C6H3(CH3)3
MDL Number MFCD00008538
Quantity 5 mL
Synonym 1,3,5-Trimethylbenzene
Molecular Weight (g/mol) 120.19 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.