Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mesitylene, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018504 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8504 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8504 Supplier MilliporeSigma Supelco Supplier No. 639085ML
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 108-67-8
Ignition Point 1022°F
Vapor Pressure 14 mmHg (55°C)
Vapor Density 4.1 (vs. air)
Density 0.864 g/mL (at 25°C)
Flash Point 50°C
Physical Form Neat
Linear Formula C6H3(CH3)3
MDL Number MFCD00008538
Melting Point -45°C
Boiling Point 163°C to 166°C
Synonym 1,3,5-Trimethylbenzene
Type Analytical Standard
Quantity 5 mL
Formula Weight 120.19
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.