Learn More
Strem, An Ascensus Company CAS# 12129-69-0. 5g. Mesitylene tungsten tricarbonyl, 98%. MFCD00015319
SDP
Catalog No. 5090113012
Encompass
CAS# 12129-69-0. 5g. Mesitylene tungsten tricarbonyl, 98%. MFCD00015319. Molecular Weight: 388.08. Molecular Formula: C9H12W(CO)3. Color/form: yellow xtl. Strem# 74-1350. http://www.strem.com/catalog/v/74-1350/
Catalog No. 50-901-13012 Supplier Strem, An Ascensus Company Supplier No. 7413505G
CAS# 12129-69-0. 5g. Mesitylene tungsten tricarbonyl, 98%. MFCD00015319. Molecular Weight: 388.08. Molecular Formula: C9H12W(CO)3. Color/form: yellow xtl. Strem# 74-1350. http://www.strem.com/catalog/v/74-1350/