Learn More
Apexbio Technology LLC Mesna 19767-45-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061681
Encompass_Preferred
Mesna chemically classified as a sulfhydryl-containing compound functions primarily as a protective adjuvant during chemotherapy with oxazaphosphorine agents (such as cyclophosphamide and ifosfamide) Its mode of action involves direct binding and detoxification of urotoxic metabolites notably acrolein thereby facilitating their conversion into stable non-toxic derivatives and mitigating associated bladder toxicity Due to this detoxifying effect Mesna is widely utilized in oncology research and clinical studies investigating chemotherapy-induced urotoxicity prevention Additionally given its structural feature as a thiol donor Mesna finds applications in experimental studies exploring its antioxidant activity and potential cytoprotective effects
Catalog No. 50-000-61681 Supplier Apexbio Technology LLC Supplier No. A84695.1
 Restricted Item
Add to Cart
Add to Cart
Mesna chemically classified as a sulfhydryl-containing compound functions primarily as a protective adjuvant during chemotherapy with oxazaphosphorine agents (such as cyclophosphamide and ifosfamide) Its mode of action involves direct binding and detoxification of urotoxic metabolites notably acrolein thereby facilitating their conversion into stable non-toxic derivatives and mitigating associated bladder toxicity Due to this detoxifying effect Mesna is widely utilized in oncology research and clinical studies investigating chemotherapy-induced urotoxicity prevention Additionally given its structural feature as a thiol donor Mesna finds applications in experimental studies exploring its antioxidant activity and potential cytoprotective effects