Learn More
FUJIFILM Biosciences mesnchymal stemcell XSFM 250ML
Catalog No. 502892144
Encompass_Preferred
PRIME-XV MSC Expansion XSFM is a serum-free and xeno-free complete medium optimized for the maintenance and expansion of purified human MSCs. This product does not contain antibiotics. PRIME-XV MSC Expansion XSFM is used in the maintenance and expansion of purified or enriched human mesenchymal stromal cells (MSCs) under serum-free and xeno-free culture conditions. This medium is ready to use. It may also be used with additional cytokine/growth factors for desired application.
Catalog No. 50-289-2144 Supplier FUJIFILM Biosciences Supplier No. 91149250ML
PRIME-XV MSC Expansion XSFM is a serum-free and xeno-free complete medium optimized for the maintenance and expansion of purified human MSCs. This product does not contain antibiotics. PRIME-XV MSC Expansion XSFM is used in the maintenance and expansion of purified or enriched human mesenchymal stromal cells (MSCs) under serum-free and xeno-free culture conditions. This medium is ready to use. It may also be used with additional cytokine/growth factors for desired application.