Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

meso-1,2,3,4-Tetrabromobutane 98.0+%, TCI America™
SDP

Catalog No. T077525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T077525G 25 g
T0775500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T077525G Supplier TCI America Supplier No. T077525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2657-67-2
Molecular Formula C4H6Br4
Molecular Weight (g/mol) 373.708
MDL Number MFCD00070471
InChI Key HGRZLIGHKHRTRE-ZXZARUISSA-N
PubChem CID 22820355
IUPAC Name (2S,3R)-1,2,3,4-tetrabromobutane
SMILES C(C(C(CBr)Br)Br)Br

Specifications

Melting Point 116°C
Color White
Boiling Point 181°C
Quantity 25 g
Formula Weight 373.71
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material meso-1,2,3,4-Tetrabromobutane
EINECSNumber (2)-2364
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.