Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

meso-2,3-Dibromosuccinic acid, 98%, Thermo Scientific™

Catalog No. AAB243820E
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
2500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB243820E 2500 g
AAB2438222 100 g
AAB2438230 250 g
AAB2438236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAB243820E Supplier Thermo Scientific Chemicals Supplier No. B243820E

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
meso-2,3-dibromosuccinic acid is used in organic synthesis & pharmaceutical intermediate.

Solubility
Soluble in alcohol and ether. Slightly soluble in water and methenyl chloride.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-36-6
Molecular Formula C4H2Br2O4
Molecular Weight (g/mol) 273.87
MDL Number MFCD00066439
InChI Key FJWGRXKOBIVTFA-XIXRPRMCSA-L
Synonym 2r,3s-2,3-dibromosuccinic acid, meso-2,3-dibromosuccinic acid, unii-d2ii9ugq9x, d2ii9ugq9x, butanedioic acid, 2,3-dibromo-, 2r,3s-rel, 2r,3s-rel-2,3-dibromosuccinic acid, 2r,3s-2,3-dibromobutanedioic acid, meso-dibromosuccinic acid, 2,3-dibromosuccinic acid, meso, 2,3-dibromosuccinic acid meso-form mi
PubChem CID 641611
IUPAC Name (2S,3R)-2,3-dibromobutanedioic acid
SMILES [O-]C(=O)[C@@H](Br)[C@@H](Br)C([O-])=O

Specifications

Melting Point ∼280°C
Quantity 2500 g
UN Number UN3261
Beilstein 1725173
Merck Index 14,3029
Solubility Information Soluble in alcohol and ether. Slightly soluble in water and methenyl chloride.
Formula Weight 275.89
Percent Purity 98%
Chemical Name or Material meso-2,3-Dibromosuccinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 208-396-9
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.