Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich meso-2,3-Dimercaptosuccinic acid

Catalog No. D78815G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
50 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D78815G 5 g
D788150G 50 g
D788125G 25 g
D78811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. D78815G Supplier Sigma Aldrich Supplier No. D78815G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

meso-2,3-Dimercaptosuccinic acid

Specifications

CAS 304-55-2
Melting Point 196°C to 198°C (dec.) (lit.)
Molecular Formula C4H6O4S2
Linear Formula HO2CCH(SH)CH(SH)CO2H
MDL Number MFCD00064799
Quantity 5 g
Synonym 2,3-Dithio-meso-tartaric acid; DIM-SA; DMS; DMSA; Succimer
Molecular Weight (g/mol) 182.22
Percent Purity ∼98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.