Learn More
Medchemexpress LLC Meso-2,6-diaminopimelic acid | 922-54-3 | MFCD09836095 | 190.20 g·mol⁻1 | C7H14N2O4 | 5 MG
SDP
Catalog No. 5000284400
Encompass_Preferred
meso-2,6-diaminopimelic acid is an amino-dicarboxylic acid used in biochemical and microbiology research. It acts as the penultimate biosynthetic precursor to L-lysine and is an important component of bacterial peptidoglycan. Supplied as a solid reagent with storage and handling documentation, it is useful for studies of cell wall synthesis and lysine metabolism.
  • Serves as the penultimate biosynthetic precursor to L-lysine.
  • Useful for studies of bacterial cell wall synthesis and peptidoglycan.
  • Supplied as a solid; white to off-white appearance.
  • Available in small research quantities suitable for analytical and biochemical assays.
  • Documentation available, including data sheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-84400 Supplier Medchemexpress LLC Supplier No. HYW4764345MG
meso-2,6-diaminopimelic acid is an amino-dicarboxylic acid used in biochemical and microbiology research. It acts as the penultimate biosynthetic precursor to L-lysine and is an important component of bacterial peptidoglycan. Supplied as a solid reagent with storage and handling documentation, it is useful for studies of cell wall synthesis and lysine metabolism.
  • Serves as the penultimate biosynthetic precursor to L-lysine.
  • Useful for studies of bacterial cell wall synthesis and peptidoglycan.
  • Supplied as a solid; white to off-white appearance.
  • Available in small research quantities suitable for analytical and biochemical assays.
  • Documentation available, including data sheet, certificate of analysis, and safety data sheet.