Learn More
Medchemexpress LLC meso-CF3-BODIPY 2 | 1642327-22-7 | 98.6% | 316.08 g/mol | 100 MG
SDP
Catalog No. 5000432010
Encompass_Preferred
meso-CF3-BODIPY 2 is a fluorescent dye that exhibits an absorption wavelength (λabs) of 553 nm and an emission wavelength (λem) of 622 nm.
  • Can be used in labeling reagents.
  • Applicable in photodynamic therapy.
  • Potential utility in organic photovoltaics and organic light-emitting diodes.
Catalog No. 50-004-32010 Supplier Medchemexpress LLC Supplier No. HYD1299100MG
meso-CF3-BODIPY 2 is a fluorescent dye that exhibits an absorption wavelength (λabs) of 553 nm and an emission wavelength (λem) of 622 nm.
  • Can be used in labeling reagents.
  • Applicable in photodynamic therapy.
  • Potential utility in organic photovoltaics and organic light-emitting diodes.