Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Meso-erythritol | 149-32-6 | MFCD00004710 | 99.7% | 122.12 g/mol | C4H10O4 | 1 ML
SDP

Catalog No. 5000211602
Encompass_Preferred

meso-Erythritol is a high-purity four-carbon sugar alcohol supplied as an analytical standard for research and analytical applications. It serves as a reference material for method development, calibration, and qualitative and quantitative analysis by HPLC, GC, and MS. The compound is provided as a white to off-white solid with purity confirmed by HPLC.

  • High purity: 99.73% (HPLC)
  • CAS number: 149-32-6
  • Molecular formula: C4H10O4; molecular weight: 122.12 g/mol
  • Intended for HPLC, GC, and MS method development and calibration
  • Physical form: white to off-white solid suitable for analytical use
  • Common pack sizes: 10 mg, 25 mg, 50 mg, 100 mg; larger sizes available by quote

Catalog No. 50-002-11602 Supplier Medchemexpress LLC Supplier No. HY10055110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

meso-Erythritol is a high-purity four-carbon sugar alcohol supplied as an analytical standard for research and analytical applications. It serves as a reference material for method development, calibration, and qualitative and quantitative analysis by HPLC, GC, and MS. The compound is provided as a white to off-white solid with purity confirmed by HPLC.

  • High purity: 99.73% (HPLC)
  • CAS number: 149-32-6
  • Molecular formula: C4H10O4; molecular weight: 122.12 g/mol
  • Intended for HPLC, GC, and MS method development and calibration
  • Physical form: white to off-white solid suitable for analytical use
  • Common pack sizes: 10 mg, 25 mg, 50 mg, 100 mg; larger sizes available by quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.