Learn More
Medchemexpress LLC Meso-erythritol | 149-32-6 | MFCD00004710 | 99.7% | 122.12 g/mol | C4H10O4 | 10 G
SDP
Catalog No. 5000211821
Encompass_Preferred
meso-Erythritol is an analytical-grade standard of the sugar alcohol erythritol supplied for research and analytical use. It serves as a reference material for method development and validation in chromatographic and mass spectrometric assays, and is appropriate for qualitative and quantitative studies.
  • Analytical standard grade with high reported purity.
  • Suitable as a reference for HPLC, GC, and MS analyses.
  • Supports qualitative and quantitative method development.
  • Available in multiple pack sizes, including 10 g.
  • Chemical identifiers: CAS 149-32-6; formula C4H10O4; molecular weight 122.12 g/mol.
Catalog No. 50-002-11821 Supplier Medchemexpress LLC Supplier No. HY10055110G
meso-Erythritol is an analytical-grade standard of the sugar alcohol erythritol supplied for research and analytical use. It serves as a reference material for method development and validation in chromatographic and mass spectrometric assays, and is appropriate for qualitative and quantitative studies.
  • Analytical standard grade with high reported purity.
  • Suitable as a reference for HPLC, GC, and MS analyses.
  • Supports qualitative and quantitative method development.
  • Available in multiple pack sizes, including 10 g.
  • Chemical identifiers: CAS 149-32-6; formula C4H10O4; molecular weight 122.12 g/mol.