Learn More
Medchemexpress LLC Meso-erythritol | 149-32-6 | MFCD00004710 | 99.7% | 122.12 g/mol | C4H10O4 | 100 MG
SDP
Catalog No. 5000211600
Encompass_Preferred
meso-Erythritol (Standard) is an analytical reference standard of the sugar alcohol erythritol intended for research and analytical applications. It occurs naturally in foods and is approximately 60-80% as sweet as sucrose. The material is supplied at high purity for use in qualitative, quantitative, and methodological experiments with chromatographic and mass spectrometric methods.
  • High purity (99.73%) analytical grade.
  • Suitable as a reference standard for HPLC, GC, and MS.
  • Useful for qualitative and quantitative method development.
  • Molecular formula C4H10O4; molecular weight 122.12 g/mol.
  • For research use only; not for patient use.
Catalog No. 50-002-11600 Supplier Medchemexpress LLC Supplier No. HY100551100G
meso-Erythritol (Standard) is an analytical reference standard of the sugar alcohol erythritol intended for research and analytical applications. It occurs naturally in foods and is approximately 60-80% as sweet as sucrose. The material is supplied at high purity for use in qualitative, quantitative, and methodological experiments with chromatographic and mass spectrometric methods.
  • High purity (99.73%) analytical grade.
  • Suitable as a reference standard for HPLC, GC, and MS.
  • Useful for qualitative and quantitative method development.
  • Molecular formula C4H10O4; molecular weight 122.12 g/mol.
  • For research use only; not for patient use.