Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Meso-erythritol | 149-32-6 | MFCD00004710 | 99.7% | 122.12 g/mol | C4H10O4 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY100551R100MG

Encompass_Preferred

An analytical reference standard of meso-erythritol intended for research and analytical use. Supplied as a high-purity sugar alcohol for qualitative and quantitative experiments, it is suitable for HPLC, GC, and mass spectrometry and is provided in small package sizes for laboratory analysis.

  • Purity 99.73%.
  • Suitable for qualitative and quantitative HPLC, GC, and MS analyses.
  • Available in 10 mg, 25 mg, 50 mg, 100 mg packages.
  • Chemical formula C4H10O4; molecular weight 122.12 g/mol.
  • Supplied with certificate of analysis for traceability.
  • Intended for research and analytical applications only.

Catalog No. 50-002-12682


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.