Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Meso-erythritol | 149-32-6 | MFCD00004710 | 99.7% | 122.12 g/mol | C4H10O4 | 25 G
SDP

Catalog No. 5000211907
Encompass_Preferred

meso-Erythritol is a high-purity analytical standard of the sugar alcohol erythritol, provided for research and analytical applications. It occurs naturally in fruits, is about 60-80% as sweet as sucrose, and is supplied as a white crystalline material with confirmed purity suitable for analytical work.

  • High purity (99.73% by HPLC).
  • Suitable as an analytical standard for research and quality control.
  • Soluble in water and DMSO.
  • Molecular weight 122.12 g/mol; molecular formula C4H10O4.
  • Applicable as a low-calorie sweetener reference and in biochemical assays.

Catalog No. 50-002-11907 Supplier Medchemexpress LLC Supplier No. HY10055125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

meso-Erythritol is a high-purity analytical standard of the sugar alcohol erythritol, provided for research and analytical applications. It occurs naturally in fruits, is about 60-80% as sweet as sucrose, and is supplied as a white crystalline material with confirmed purity suitable for analytical work.

  • High purity (99.73% by HPLC).
  • Suitable as an analytical standard for research and quality control.
  • Soluble in water and DMSO.
  • Molecular weight 122.12 g/mol; molecular formula C4H10O4.
  • Applicable as a low-calorie sweetener reference and in biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.