Learn More
Medchemexpress LLC Meso-erythritol | 149-32-6 | MFCD00004710 | 99.7% | 122.12 g/mol | C4H10O4 | 25 MG
SDP
Catalog No. 5000213733
Encompass_Preferred
Analytical standard of meso-erythritol supplied as a 25 mg reference material for research and analytical applications. This sugar alcohol is commonly used in method development, calibration, and qualitative or quantitative analysis by HPLC, GC, and MS. Certificate of analysis is provided; for research use only.
  • High purity: 99.73%.
  • Compact pack size: 25 MG.
  • Known identifiers: CAS 149-32-6, molecular weight 122.12 g/mol.
  • Suitable for HPLC, GC, and MS method development.
  • Provided with certificate of analysis for traceability.
  • Intended for research use only; follow CoA storage instructions.
Catalog No. 50-002-13733 Supplier Medchemexpress LLC Supplier No. HY100551R25MG
Analytical standard of meso-erythritol supplied as a 25 mg reference material for research and analytical applications. This sugar alcohol is commonly used in method development, calibration, and qualitative or quantitative analysis by HPLC, GC, and MS. Certificate of analysis is provided; for research use only.
  • High purity: 99.73%.
  • Compact pack size: 25 MG.
  • Known identifiers: CAS 149-32-6, molecular weight 122.12 g/mol.
  • Suitable for HPLC, GC, and MS method development.
  • Provided with certificate of analysis for traceability.
  • Intended for research use only; follow CoA storage instructions.