Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Meso-erythritol | 149-32-6 | MFCD00004710 | 99.7% | 122.12 g/mol | C4H10O4 | 5 G
SDP

Catalog No. 5000213732
Encompass_Preferred

meso-erythritol (CAS 149-32-6) is a four-carbon sugar alcohol supplied as a white to off-white solid for research and analytical use. It serves as a low-calorie sweetener reference and an analytical standard in biochemical and chromatography applications, with high reported purity suitable for laboratory studies.

  • High purity: 99.7%.
  • White to off-white solid appearance.
  • Chemical formula C4H10O4 and molecular weight 122.12 g/mol.
  • Supplied in small laboratory sizes including a 5 g package.
  • Intended for analytical standards and research applications.

Catalog No. 50-002-13732 Supplier Medchemexpress LLC Supplier No. HY1005515G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

meso-erythritol (CAS 149-32-6) is a four-carbon sugar alcohol supplied as a white to off-white solid for research and analytical use. It serves as a low-calorie sweetener reference and an analytical standard in biochemical and chromatography applications, with high reported purity suitable for laboratory studies.

  • High purity: 99.7%.
  • White to off-white solid appearance.
  • Chemical formula C4H10O4 and molecular weight 122.12 g/mol.
  • Supplied in small laboratory sizes including a 5 g package.
  • Intended for analytical standards and research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.