Learn More
Medchemexpress LLC Meso-erythritol | 149-32-6 | MFCD00004710 | 99.7% | 122.12 g/mol | C4H10O4 | 50 G
SDP
Catalog No. 5000211818
Encompass_Preferred
meso-Erythritol (Standard) is an analytical reference standard of the sugar alcohol meso-erythritol (CAS 149-32-6). It is a solid, high-purity compound intended for research and analytical applications, commonly used as a reference in chromatographic and mass-spectrometric methods for qualitative and quantitative analysis.
Catalog No. 50-002-11818 Supplier Medchemexpress LLC Supplier No. HY10055150G
meso-Erythritol (Standard) is an analytical reference standard of the sugar alcohol meso-erythritol (CAS 149-32-6). It is a solid, high-purity compound intended for research and analytical applications, commonly used as a reference in chromatographic and mass-spectrometric methods for qualitative and quantitative analysis.