Learn More
Medchemexpress LLC Meso-erythritol | 149-32-6 | MFCD00004710 | 99.7% | 122.12 g/mol | C4H10O4 | 500 G
SDP
Catalog No. 5000212707
Encompass_Preferred
meso-Erythritol is a high-purity sugar alcohol used as a low-calorie sweetener and food additive. It naturally occurs in fruits such as pear and watermelon and provides about 60-80% of the sweetness of sucrose while contributing fewer calories.
  • High purity (99.73%) suitable for research and formulation
  • Approximately 60-80% as sweet as sucrose
  • Water-soluble (≥100 mg/mL), facilitating aqueous formulations
  • Low-calorie alternative with favorable digestive tolerance
  • Available in multiple pack sizes including 500 G
Catalog No. 50-002-12707 Supplier Medchemexpress LLC Supplier No. HY100551500G
meso-Erythritol is a high-purity sugar alcohol used as a low-calorie sweetener and food additive. It naturally occurs in fruits such as pear and watermelon and provides about 60-80% of the sweetness of sucrose while contributing fewer calories.
  • High purity (99.73%) suitable for research and formulation
  • Approximately 60-80% as sweet as sucrose
  • Water-soluble (≥100 mg/mL), facilitating aqueous formulations
  • Low-calorie alternative with favorable digestive tolerance
  • Available in multiple pack sizes including 500 G