Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

meso-Hydrobenzoin 98.0+%, TCI America™
SDP

Catalog No. H081625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H081625G 25 g
H08165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H081625G Supplier TCI America Supplier No. H081625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 579-43-1
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00064253
InChI Key IHPDTPWNFBQHEB-UHFFFAOYNA-N
Synonym meso-hydrobenzoin, meso-1,2-diphenyl-1,2-ethanediol, unii-co9a49a84i, meso-stilbene glycol, 1r,2s-1,2-diphenylethane-1,2-diol, meso-1,2-diphenylethylene glycol, hydrobenzoin, meso, unii-q61g3433lb component
PubChem CID 853018
ChEBI CHEBI:50015
IUPAC Name 1,2-diphenylethane-1,2-diol
SMILES OC(C(O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 136°C
Color White
Quantity 25 g
Formula Weight 214.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material meso-Hydrobenzoin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.