Learn More
Strem, An Ascensus Company CAS# 14609-54-2. 1g. meso-Tetra(4-carboxyphenyl)porphine, 98%. MFCD00064860
SDP
Catalog No. 5090109119
Encompass
CAS# 14609-54-2. 1g. meso-Tetra(4-carboxyphenyl)porphine, 98%. MFCD00064860. Molecular Weight: 790.79. Molecular Formula: C48H30N4O8. Color/form: purple pwdr. Strem# 07-1970. http://www.strem.com/catalog/v/07-1970/
Catalog No. 50-901-09119 Supplier Strem, An Ascensus Company Supplier No. 0719701G
CAS# 14609-54-2. 1g. meso-Tetra(4-carboxyphenyl)porphine, 98%. MFCD00064860. Molecular Weight: 790.79. Molecular Formula: C48H30N4O8. Color/form: purple pwdr. Strem# 07-1970. http://www.strem.com/catalog/v/07-1970/