Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Meso-tetrakis(4-aminophenyl)porphyrin | 22112-84-1 | MFCD00191497 | >97.0% | 674.79 g/mol | C44H34N8 | 5 G
SDP

Catalog No. 5000299349
Encompass_Preferred

Meso-tetrakis(4-aminophenyl)porphyrin is an amine-substituted tetraphenylporphyrin used in research applications such as porphyrin synthesis, photophysics, and as a functional building block in materials and catalysis. It is commonly handled as a solid research reagent and characterized by CAS 22112-84-1 and a molecular weight of 674.79 g/mol.

  • Amine-functionalized porphyrin core suitable for derivatization.
  • Molecular formula C44H34N8 and molecular weight 674.79 g/mol.
  • Useful for photophysical, catalytic, and materials chemistry studies.
  • Available in small research pack sizes appropriate for laboratory work.
  • Storage: store at 4°C, protect from light; in solution, -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-99349 Supplier Medchemexpress LLC Supplier No. HYW0351365G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Meso-tetrakis(4-aminophenyl)porphyrin is an amine-substituted tetraphenylporphyrin used in research applications such as porphyrin synthesis, photophysics, and as a functional building block in materials and catalysis. It is commonly handled as a solid research reagent and characterized by CAS 22112-84-1 and a molecular weight of 674.79 g/mol.

  • Amine-functionalized porphyrin core suitable for derivatization.
  • Molecular formula C44H34N8 and molecular weight 674.79 g/mol.
  • Useful for photophysical, catalytic, and materials chemistry studies.
  • Available in small research pack sizes appropriate for laboratory work.
  • Storage: store at 4°C, protect from light; in solution, -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.