Learn More
Medchemexpress LLC Meso-tetrakis(4-aminophenyl)porphyrin | 22112-84-1 | MFCD00191497 | 98.0% | 674.79 g·mol⁻¹ | C44H34N8 | 250 MG
SDP
Catalog No. 5000303502
Encompass_Preferred
Meso-tetrakis(4-aminophenyl)porphyrin is a porphyrin derivative supplied as a high-purity solid for research and analytical applications. The compound has molecular formula C44H34N8, molecular weight 674.79 g·mol⁻¹, and CAS 22112-84-1. It should be protected from light and stored under inert atmosphere for stability.
  • High purity (98.0%) suitable for research use.
  • Distinct deep purple to black solid for easy visual identification.
  • Defined chemical composition: C44H34N8, 674.79 g·mol⁻¹.
  • Recommended storage: solid at 4°C, protected from light and under nitrogen.
  • Solution stability: -80°C up to 6 months, -20°C up to 1 month with light protection.
  • Available in multiple pack sizes including 250 mg for flexible sampling.
Catalog No. 50-003-03502 Supplier Medchemexpress LLC Supplier No. 250MGHYW035136
Add to Cart
Add to Cart
Meso-tetrakis(4-aminophenyl)porphyrin is a porphyrin derivative supplied as a high-purity solid for research and analytical applications. The compound has molecular formula C44H34N8, molecular weight 674.79 g·mol⁻¹, and CAS 22112-84-1. It should be protected from light and stored under inert atmosphere for stability.
  • High purity (98.0%) suitable for research use.
  • Distinct deep purple to black solid for easy visual identification.
  • Defined chemical composition: C44H34N8, 674.79 g·mol⁻¹.
  • Recommended storage: solid at 4°C, protected from light and under nitrogen.
  • Solution stability: -80°C up to 6 months, -20°C up to 1 month with light protection.
  • Available in multiple pack sizes including 250 mg for flexible sampling.