Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Meso-tetrakis(4-aminophenyl)porphyrin | 22112-84-1 | MFCD00191497 | ≥98.0% | 674.79 g·mol⁻¹ | C44H34N8 | 500 MG
SDP

Catalog No. 5000299298
Encompass_Preferred

Meso-tetrakis(4-aminophenyl)porphyrin is a porphyrin derivative used as a research reagent and building block in porphyrin chemistry, coordination complexes, materials science, and organic synthesis. The material is supplied as a high-purity solid suitable for laboratory use.

  • High purity: ≥98.0% (NMR).
  • Molecular weight: 674.79 g·mol⁻¹.
  • Chemical formula: C44H34N8.
  • Appearance: deep purple to black solid.
  • Suitable for synthesis and materials research.
  • Characterized by a manufacturer certificate of analysis.

Catalog No. 50-002-99298 Supplier Medchemexpress LLC Supplier No. HYW035136500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Meso-tetrakis(4-aminophenyl)porphyrin is a porphyrin derivative used as a research reagent and building block in porphyrin chemistry, coordination complexes, materials science, and organic synthesis. The material is supplied as a high-purity solid suitable for laboratory use.

  • High purity: ≥98.0% (NMR).
  • Molecular weight: 674.79 g·mol⁻¹.
  • Chemical formula: C44H34N8.
  • Appearance: deep purple to black solid.
  • Suitable for synthesis and materials research.
  • Characterized by a manufacturer certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.