Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

meso-Tetraphenylporphine, 97%

Catalog No. p-199292
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC171210010 1 g Glass bottle
AC171210050 5 g Glass bottle
AC171210250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC171210010 Supplier Thermo Scientific Chemicals Supplier No. 171210010
Only null left
Add to Cart

CAS: 917-23-7 | C44H30N4 | 614.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 917-23-7
Molecular Formula C44H30N4
Molecular Weight (g/mol) 614.75
MDL Number MFCD00011680
InChI Key AQPPOLXYUQPDOD-UHFFFAOYSA-N
Synonym tetraphenylporphyrin, 5,10,15,20-tetraphenylporphyrin, meso-tetraphenylporphine, tetraphenylporphine, 5,10,15,20-tetraphenylporphine, meso-tetraphenylporphyrin, 21h,23h-porphine, 5,10,15,20-tetraphenyl, 5,10,15,20-tetraphenyl-21h,23h-porphine, meso-tetraphenylporphyrine, tetraphenyl porphine
PubChem CID 70186
IUPAC Name 5,10,15,20-tetraphenyl-21,22-dihydroporphyrin
SMILES N1C2=CC=C1C(=C1C=CC(=N1)C(=C1C=CC(=N1)C(=C1NC(C=C1)=C2C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point >300.0°C
Color Blue to Purple
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in alcohol
Formula Weight 614.75
Percent Purity 97%
Physical Form Glistening Crystalline Powder
Chemical Name or Material meso-Tetraphenylporphine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
W
EINECSNumber 213-025-9
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.