Learn More
Cayman Chemical meso2 3DImercaptosuccInIc A 5g
SDP
Catalog No. 502940058
Encompass_Preferred
A heavy metal chelator; reverses sodium arsenite-induced inhibition of PDH in mouse kidney homogenates (ED50 = 0.25 mM); decreases cisplatin-induced increases in serum levels of platinum and BUN, as well as apoptosis in renal tubules of mice at 100 µM/kg; decreases elevations in the activity of catalase and G6PDH in RBCs from lead exposed rats at 90 mg/kg; decreases the levels of lead in the cerebellum, hippocampus, liver, kidney, and blood in rats at 100 mg/kg; decreases immobility time in the forced swim test in lead-exposed male, but not female, mice at 50 mg/kg
Catalog No. 50-294-0058 Supplier Cayman Chemical Supplier No. 285155 G
A heavy metal chelator; reverses sodium arsenite-induced inhibition of PDH in mouse kidney homogenates (ED50 = 0.25 mM); decreases cisplatin-induced increases in serum levels of platinum and BUN, as well as apoptosis in renal tubules of mice at 100 µM/kg; decreases elevations in the activity of catalase and G6PDH in RBCs from lead exposed rats at 90 mg/kg; decreases the levels of lead in the cerebellum, hippocampus, liver, kidney, and blood in rats at 100 mg/kg; decreases immobility time in the forced swim test in lead-exposed male, but not female, mice at 50 mg/kg