Learn More
Cayman Chemical Mesoridazine, Purity: ≥95%, CAS: 5588-33-0, 100 mg
SDP
Catalog No. 502991883
Encompass_Preferred
An active metabolite of thioridazine; formed via sulfoxidation of thioridazine by CYP2D6 in human liver microsomes; binds to histamine H1, dopamine D2, muscarinic acetylcholine, and α1- and α2-adrenergic receptors (Kds = 1.8, 19, 69, 2, and 1,600 nM, respectively); prevents mescaline-induced hyperactivity in mice at 10, 30, and 45 mg/kg
Catalog No. 50-299-1883 Supplier Cayman Chemical Supplier No. 31370100 MG
An active metabolite of thioridazine; formed via sulfoxidation of thioridazine by CYP2D6 in human liver microsomes; binds to histamine H1, dopamine D2, muscarinic acetylcholine, and α1- and α2-adrenergic receptors (Kds = 1.8, 19, 69, 2, and 1,600 nM, respectively); prevents mescaline-induced hyperactivity in mice at 10, 30, and 45 mg/kg