Learn More
Cayman Chemical MesorIdazIn 10mg
SDP
Catalog No. 502990689
Encompass_Preferred
An active metabolite of thioridazine; formed via sulfoxidation of thioridazine by CYP2D6 in human liver microsomes; binds to histamine H1, dopamine D2, muscarinic acetylcholine, and α1- and α2-adrenergic receptors (Kds = 1.8, 19, 69, 2, and 1,600 nM, respectively); prevents mescaline-induced hyperactivity in mice at 10, 30, and 45 mg/kg
Catalog No. 50-299-0689 Supplier Cayman Chemical Supplier No. 3137010 MG
An active metabolite of thioridazine; formed via sulfoxidation of thioridazine by CYP2D6 in human liver microsomes; binds to histamine H1, dopamine D2, muscarinic acetylcholine, and α1- and α2-adrenergic receptors (Kds = 1.8, 19, 69, 2, and 1,600 nM, respectively); prevents mescaline-induced hyperactivity in mice at 10, 30, and 45 mg/kg