Learn More
TARGETMOL CHEMICALS INC Mesoridazine Besylate 10MG
SDP
Catalog No. 502877844
Encompass_Preferred
Also available in 1 mL, 5 mg and bulk. Please contact Fisher for quotes.Mesoridazine Besylate (Serentil) is a piperidine neuroleptic drug belonging to the class of drugs called phenothiazines, used in the treatment of schizophrenia. It is a metabolite of thioridazine. Purity 99.58%
Catalog No. 50-287-7844 Supplier TARGETMOL CHEMICALS INC Supplier No. T308610MG
Also available in 1 mL, 5 mg and bulk. Please contact Fisher for quotes.Mesoridazine Besylate (Serentil) is a piperidine neuroleptic drug belonging to the class of drugs called phenothiazines, used in the treatment of schizophrenia. It is a metabolite of thioridazine. Purity 99.58%