Learn More
Apexbio Technology LLC Mesoridazine Besylate 32672-69-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061670
Encompass_Preferred
Mesoridazine besylate (CAS 32672-69-8) is a phenothiazine derivative that functions primarily as a dopamine receptor antagonist demonstrating notable inhibitory activity against dopamine D2 and D4 receptors The molecule has particularly high affinity toward the dopamine D4 receptor subtype By modulating dopamine receptor activity Mesoridazine besylate has potential relevance in neuroscience research aimed at understanding dopamine-mediated neurological pathways and disorders Frequently utilized in studies of psychotic conditions the compound serves as an important tool for investigating receptor-specific pharmacological mechanisms
Catalog No. 50-000-61670 Supplier Apexbio Technology LLC Supplier No. A87015.1
Mesoridazine besylate (CAS 32672-69-8) is a phenothiazine derivative that functions primarily as a dopamine receptor antagonist demonstrating notable inhibitory activity against dopamine D2 and D4 receptors The molecule has particularly high affinity toward the dopamine D4 receptor subtype By modulating dopamine receptor activity Mesoridazine besylate has potential relevance in neuroscience research aimed at understanding dopamine-mediated neurological pathways and disorders Frequently utilized in studies of psychotic conditions the compound serves as an important tool for investigating receptor-specific pharmacological mechanisms