Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mesosulfuron-methyl | 208465-21-8 | 98.0% | 10 G
SDP

Catalog No. 5000338697
Encompass_Preferred

Mesosulfuron-methyl is a sulfonylurea herbicide that inhibits acetolactate synthase (ALS) and is used in research for post-emergence control of ryegrass and *Avena spp.* (wild oat) in wheat fields.

  • Inhibits acetolactate synthase (ALS)
  • Used for post-emergence control of ryegrass and *Avena spp.* (wild oat) in wheat fields
  • Molecular weight: 503.51
  • Appearance: White to off-white powder
  • For research use only

Catalog No. 50-003-38697 Supplier Medchemexpress LLC Supplier No. HY12698710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mesosulfuron-methyl is a sulfonylurea herbicide that inhibits acetolactate synthase (ALS) and is used in research for post-emergence control of ryegrass and *Avena spp.* (wild oat) in wheat fields.

  • Inhibits acetolactate synthase (ALS)
  • Used for post-emergence control of ryegrass and *Avena spp.* (wild oat) in wheat fields
  • Molecular weight: 503.51
  • Appearance: White to off-white powder
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.