Learn More
Medchemexpress LLC Mesosulfuron-methyl | 208465-21-8 | 98.0% | 250 MG
SDP
Catalog No. 5000342476
Encompass_Preferred
Mesosulfuron-methyl is a sulfonylurea herbicide that functions by inhibiting acetolactate synthase (ALS), and it is used in research for post-emergence control of ryegrass and *Avena spp.* (wild oat) in wheat fields.
  • It is a sulfonylurea herbicide.
  • It inhibits acetolactate synthase (ALS).
  • It is used for post-emergence control.
  • It targets ryegrass and *Avena spp.* (wild oat).
  • Its application is in wheat fields.
  • It is intended for research use only.
Catalog No. 50-003-42476 Supplier Medchemexpress LLC Supplier No. 250MGHY126987
Mesosulfuron-methyl is a sulfonylurea herbicide that functions by inhibiting acetolactate synthase (ALS), and it is used in research for post-emergence control of ryegrass and *Avena spp.* (wild oat) in wheat fields.
  • It is a sulfonylurea herbicide.
  • It inhibits acetolactate synthase (ALS).
  • It is used for post-emergence control.
  • It targets ryegrass and *Avena spp.* (wild oat).
  • Its application is in wheat fields.
  • It is intended for research use only.